EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
[CuBr(PPh3)3] for azide-alkyne cycloaddition reactions under strict Click conditions.
PMID 21384852
Abstract
A careful methodological study revealed a true Click catalytic system based on commercially available [CuBr(PPh(3))(3)]. This system is active at room temperature, with 0.5 mol % [Cu] (or less), in the absence of any additive, and it does not require any purification step to isolate pure triazoles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G4168
α-D-Mannopyranosyl azide tetraacetate, ≥90% (TLC)
C14H19N3O9