EMAIL THIS PAGE TO A FRIEND
British journal of cancer
Epstein-Barr Virus latent membrane protein 1 induces Snail and epithelial-mesenchymal transition in metastatic nasopharyngeal carcinoma.
PMID 21386845
Abstract
Epstein-Barr Virus (EBV)-associated nasopharyngeal carcinoma (NPC) is distinctive among head-and-neck cancers in its undifferentiated histopathology and highly metastatic character. We have recently investigated the involvement of epithelial-mesenchymal transition (EMT) in NPC. In a previous study, we found a close association of expression of LMP1, the principal EBV oncoprotein, with expression of Twist and induction of EMT. We analysed expression of Snail in 41 NPC tissues by immunohistochemistry. The role of Twist as well as Snail in EMT of NPC was investigated by using NP69SV40T human nasopharyngeal cells. In NPC tissues, overexpression of Snail is associated with expression of LMP1 in carcinomatous cells. In addition, expression of Snail positively correlated with metastasis and independently correlated inversely with expression of E-cadherin. Expression of Twist had no association with expression of E-cadherin. Further, in a human nasopharyngeal cell line, LMP1 induces EMT and its associated cellular motility and invasiveness. Expression of Snail is induced by LMP1 in these cells, and small hairpin RNA (shRNA) to Snail reversed the cellular changes. By contrast, Twist did not produce EMT in these nasopharyngeal cells. This study strengthens the association of EMT with the metastatic behaviour of NPC. These results suggest that induction of Snail by the EBV oncoprotein LMP1 has a pivotal role in EMT in NPC.