EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A readily synthesized cyclic pyrrolysine analogue for site-specific protein "click" labeling.
PMID 21387054
Abstract
A concise route was developed for the facile synthesis of a cyclic pyrrolysine analogue bearing an azide handle. Directed evolution enabled the encoding of this non-natural amino acid in both prokaryotic and eukaryotic cells, which offers a highly efficient approach for the site-specific protein labeling using click chemistry.
Related Materials