EMAIL THIS PAGE TO A FRIEND
International journal of cancer
E-cadherin expression on human carcinoma cell affects trastuzumab-mediated antibody-dependent cellular cytotoxicity through killer cell lectin-like receptor G1 on natural killer cells.
PMID 21387286
Abstract
Trastuzumab is a recombinant antibody drug that is widely used for the treatment of HER2-overexpressing breast carcinoma. Despite encouraging clinical results, many HER2-overexpressing carcinomas are primarily resistant to trastuzumab. We attempted to explain trastuzumab resistance and search for solutions. Since the killer cell lectin-like receptor G1 (KLRG1), an inhibitory receptor expressed on subsets of natural killer (NK) cells recognizes E-cadherin as ligands and may inhibit immune responses by regulating the effector function of NK cells, we used HER2-overexpressing carcinoma cells which were expressing E-cadherin to investigate the role of antibody-dependent cellular cytotoxicity (ADCC) through KLRG1 on NK cells in vitro and vivo. The results indicated that HER2-overexpressing carcinoma cells were killed by trastuzumab-mediated ADCC and the ADCC activity was reflected the degree of E-cadherin expression on carcinoma cells. We found that expression of E-cadherin was shown to be a predictor of response to trastuzumab-based treatment for HER2-overexpressing carcinomas, furthermore, trastuzumab-mediated ADCC was markedly enhanced by KLRG1-negative peripheral blood mononuclear cells (PBMCs(KLRG1(-))).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
5085 E-Cadherin human, recombinant, expressed in E. coli, 0.5 mg protein/mL