EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Sequential one-pot ruthenium-catalyzed azide-alkyne cycloaddition from primary alkyl halides and sodium azide.
PMID 21388208
Abstract
An experimentally simple sequential one-pot RuAAC reaction, affording 1,5-disubstituted 1H-1,2,3-triazoles in good to excellent yields starting from an alkyl halide, sodium azide, and an alkyne, is reported. The organic azide is formed in situ by treating the primary alkyl halide with sodium azide in DMA under microwave heating. Subsequent addition of [RuClCp*(PPh(3))(2)] and the alkyne yielded the desired cycloaddition product after further microwave irradiation.