EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Transplantation of a mammary stromal cell line into a mammary fat pad: development of the site-specific in vivo analysis system for mammary stromal cells.
PMID 21389616
Abstract
The interaction between mammary epithelial and stromal tissue is considered to be important in breast tissue development. In this study, we developed a transplantation procedure for the mammary stromal fibroblastic cell line (MSF) to examine its life in vivo. First we established MSF cells which stably expressed lacZ (lacZ/MSF) and had characteristics of mammary stromal cells. The lacZ/MSF cells were then transplanted into a cleared mammary fat pad of syngenic mice with and without mammary primary epithelial organoids. Whole mount X-gal and carmine staining of the transplants revealed that a number of undifferentiated lacZ/MSF cells survived around the mammary epithelial tissue when transplanted with organoids. These results indicate that transplantation of MSF cells into mammary fat pad was accomplished by co-transplantation with primary mammary organoids. Finally, we discuss the application of transplantation procedure for in vivo studies of the mammary stromal tissue development and stromal-epithelial interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
229253
Carminic acid
C22H20O13
11298
Carminic acid, analytical standard
C22H20O13