EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section F, Structural biology and crystallization communications
Crystallization and preliminary X-ray crystallographic study of a methyltransferase involved in 2-methylisoborneol biosynthesis in Streptomyces lasaliensis.
PMID 21393856
Abstract
The biosynthetic pathway of the off-flavour terpenoid alcohol 2-methylisoborneol (2-MIB) requires geranyl pyrophosphate methyltransferase (GPPMT) to methylate GPP before the cyclization reaction. GPPMT is the first example of an S-adenosyl-L-methionine-dependent methyltransferase that acts on general intermediates such as geranyl pyrophosphate and farnesyl pyrophosphate in isoprenoid biosynthetic pathways. In this study, recombinant GPPMT was overproduced, purified and crystallized in the absence and presence of cofactor, cofactor analogue and substrate. Well diffracting crystals of apo GPPMT containing one molecule in the asymmetric unit were obtained and the structure of this form was solved by the molecular-replacement method. Two crystal forms of the tertiary complex with GPP and sinefungin were also obtained. Structure analysis of these crystals is currently under way in order to understand the enzyme reaction mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76532
Geranyl pyrophosphate lithium salt, ≥95.0% (TLC)
C10H20O7P2 · xLi+
19533
Geranyl pyrophosphate lithium salt, analytical standard
C10H20O7P2 · xLi+