EMAIL THIS PAGE TO A FRIEND
ChemSusChem
Direct catalytic synthesis of 5-methylfurfural from biomass-derived carbohydrates.
PMID 21394924
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W270202
5-Methylfurfural, ≥98%, FG
C6H6O2
137316
5-Methylfurfural, ReagentPlus®, 99%
C6H6O2