EMAIL THIS PAGE TO A FRIEND
Vestnik Rossiiskoi akademii meditsinskikh nauk
[Effectiveness of duspatalin therapy in the treatment and prevention of post-cholecystectomy syndrome].
PMID 21395089
Abstract
Results of evaluation of the efficiency of myotropic spasmolytic Duspatalin during long-term therapy and preventive treatment of functional post-cholecystectomy syndrome are presented. The influence of the treatment on manifestations of clinical symptoms, quality of a life estimated based on a visual-analog scale, and intestinal microbiocenosis (changes in the activity of short-chain fatty acids) are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3282
Mebeverine hydrochloride, analytical standard
C25H35NO5 · HCl