EMAIL THIS PAGE TO A FRIEND
Behavioural brain research
Triadimefon supports conditioned cue preference.
PMID 21396967
Abstract
Triadimefon (TDF) is a fungicide with effects similar to cocaine, suggesting potential for abuse. Mice were trained in an apparatus with two distinctive flooring cues. In the experimental group, one of these was paired with administration of TDF and the other with vehicle; in the control group, both flooring types were paired with vehicle. The experimental group showed a significant preference shift towards the TDF-paired cue, suggesting a reinforcing effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45693
Triadimefon, PESTANAL®, analytical standard
C14H16ClN3O2
442818
Triadimefon, analytical standard
C14H16ClN3O2