EMAIL THIS PAGE TO A FRIEND
International journal of obstetric anesthesia
Estimation of infant dose and exposure to pethidine and norpethidine via breast milk following patient-controlled epidural pethidine for analgesia post caesarean delivery.
PMID 21398109
Abstract
There is no information about the distribution of pethidine into breast milk and/or exposure of the breastfed infant during pethidine patient-controlled epidural analgesia after caesarean delivery. We conducted an observational study among 20 women. The mean (95% confidence interval) pethidine dose administered was 670 (346-818) mg over 41 (35-46) h. Maternal plasma and milk and neonatal plasma were collected near the time of pethidine cessation and 6h later. Absolute and relative infant doses via milk and infant exposure were calculated. Infant behaviour was assessed using the Neurologic and Adaptive Capacity Score. At first and second sampling times, mean absolute infant doses for pethidine were 20 (14-27) μg/kg/day and 10 (7-13) μg/kg/day, while mean relative infant doses were 0.7 (0.1-1.4)% and 0.3 (0.1-0.5)% respectively. Similar values for norpethidine (expressed as pethidine equivalents) were 21 (16-26) μg/kg/day and 22 (12-32) μg/kg/day; and 0.7 (0.3-1)% and 0.6 (0.2-1)% respectively. Mean pethidine and norpethidine concentrations in neonatal plasma were 3 (0-6.1) μg/L and 0.6 (0.2-1) μg/L. Compared with a time-matched maternal sample, the infant's exposure was 1.4 (0.2-2.8)% for pethidine and 0.4 (0.2-0.6)% for norpethidine. The mean (95% confidence interval) neurologic and adaptive capacity score was 33.6 (32.2-34.9). The combined absolute infant dose of pethidine and norpethidine received via milk was 1.8% of the neonatal therapeutic dose and the combined relative infant dose was below the 10% recommended safety level. Breastfed infants are at low risk of drug exposure when mothers self-administer epidural pethidine after caesarean delivery.
Related Materials