EMAIL THIS PAGE TO A FRIEND
Physiological research
Short-term regulation of adiponectin secretion in rat adipocytes.
PMID 21401303
Abstract
Adiponectin belongs to the group of biologically active substances secreted by adipocytes and referred to as adipokines. Disturbances in its secretion and/or action are thought to be involved in the pathogenesis of some metabolic diseases. However, regulation of adiponectin secretion is poorly elucidated. In the present study, short-term regulation of adiponectin secretion in primary rat adipocytes was investigated. Isolated rat adipocytes were incubated in Krebs-Ringer buffer containing 5 mM glucose and insulin alone or in the combination with epinephrine, dibutyryl-cAMP, adenosine A(1) receptor antagonist (DPCPX), palmitate, 2-bromopalmitate or inhibitor of mitochondrial electron transport (rotenone). Adipocyte exposure for 2 h to insulin (1-100 nM) significantly increased secretion of adiponectin compared with secretion observed without insulin. Furthermore, secretion of adiponectin from adipocytes incubated with glucose and insulin was reduced by 1 and 2 microM epinephrine, but not by 0.25 and 0.5 microM epinephrine. Under similar conditions, 1 and 2 mM dibutyryl-cAMP substantially diminished secretion of adiponectin, whereas 0.5 mM dibutyryl-cAMP was ineffective. Secretion of adiponectin was found to be effectively decreased by DPCPX. Moreover, adipocyte exposure to rotenone also resulted in a substantial diminution of secretory response of adipocytes incubated for 2 h with glucose and insulin. It was also demonstrated that palmitate and 2-bromopalmitate (0.06-0.5 mM) failed to affect secretion of leptin. The obtained results indicated that in short-term regulation of adiponectin secretion, insulin and epinephrine exert the opposite effects. These effects appeared as early as after 2 h of exposure. Moreover, deprivation of energy or blockade of adenosine action substantially decreased secretion of adiponectin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
21604
2-Bromohexadecanoic acid, ≥99.0% (GC)
C16H31BrO2
238422
2-Bromohexadecanoic acid, ~97%
C16H31BrO2