EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Changes in antibiotic distribution due to pancreatitis.
PMID 21402839
Abstract
This work sought to define how pancreatitis affected antibiotic distribution in a perfused rat pancreas model. The distribution kinetics of four antibiotics were examined in control animals and animals with pancreatitis. Meropenem and piperacillin distributed into the extracellular space, and their distribution kinetics were unaffected by pancreatitis. In contrast, in pancreatic cells from animals with pancreatitis, ciprofloxacin showed a reduced uptake and clindamycin showed a reduced distribution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
205494
Dibutyltin dichloride, 96%
C8H18Cl2Sn