EMAIL THIS PAGE TO A FRIEND
Archives of ophthalmology (Chicago, Ill. : 1960)
Common features of periocular tinea.
PMID 21402986
Abstract
To present the common features of periocular tinea to aid physicians in future diagnosis and therapy of this condition, because superficial fungal infections on the face are often misdiagnosed owing to the diverse morphologies that they manifest. This is especially true of dermatophytoses involving the periocular region. A retrospective review was performed of patients with a diagnosis of periocular tinea who were seen between January 2003 and September 2009 in the pediatric dermatology clinic at St. Louis Children's Hospital. Ten cases of periocular tinea were identified (6 male patients and 4 female patients). Common features included prolonged misdiagnosis (all 10 cases), a normal ophthalmologic examination (all 10 cases), and inappropriate corticosteroid application (7 cases). Loss of the eyelashes occurred in all 10 patients. No cases had evidence of other tinea infections on examination. Only 2 cases had the central clearing classically associated with tinea corporis. Seven patients had a potassium hydroxide preparation and/or culture positive for fungal elements. Lesions improved with topical and oral antifungal treatment in all cases, and patients were able to regrow their eyelashes. Periocular tinea should be considered in the differential diagnosis for periocular inflammation, especially in those patients refractory to therapy for more common conditions. Loss of the eyelashes is characteristic of these fungal infections, similar to the hair loss that occurs in kerions associated with tinea capitis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E0050000 Econazole nitrate, European Pharmacopoeia (EP) Reference Standard
C18H16Cl3N3O4