EMAIL THIS PAGE TO A FRIEND
Huan jing ke xue= Huanjing kexue
[Migration of main odorous compounds in a water supply system with Huangpu River as raw water in Shanghai].
PMID 21404674
Abstract
Migration and variation of odorous compounds as geosmin, 2-methylisoborneol (MIB) and residual chlorine in drinking water taken from Huangpu River were studied by using headspace solid phase microextraction procedure (HSPME) and gas chromatograph with mass spectrometry. The results showed that, raw water, processed water, pipe water (taken from pump station) and secondary-supply water all contained MIB and geosmin ranging from 2 ng/L to 18 ng/L and 2.68 ng/L to 5.06 ng/L respectively and decreased dramatically during the water processing and distribution system. MIB is proved to be a kind of the odorous compounds in drinking water of Shanghai by comparing the concentrations of MIB and GSM with their odor threshold. The concentration of residual chlorine declined greatly in the distribution system, but because of the high value at the outlet of waterworks, it still may exceeded the influence of MIB and cause the odour problems.
Related Materials