EMAIL THIS PAGE TO A FRIEND
Inorganic chemistry
Peculiarities of the complexation of copper and silver adducts of a 3,5-bis(trifluoromethyl)pyrazolate ligand with organoiron compounds.
PMID 21405060
Abstract
Interaction of the copper, {[3,5-(CF(3))(2)Pz]Cu}(3), and silver, {[3,5-(CF(3))(2)Pz]Ag}(3), macrocycles [3,5-(CF(3))(2)Pz = 3,5-bis(trifluoromethyl)pyrazolate] with cyclooctatetraeneiron tricarbonyl, (cot)Fe(CO)(3), was investigated by IR and NMR spectroscopy for the first time. The formation of 1:1 complexes was observed at low temperatures in hexane. The composition of the complexes (1:1) and their thermodynamic characteristics in hexane and dichloromethane were determined. The π-electron system of (cot)Fe(CO)(3) was proven to be the sole site of coordination in solution and in the solid state. However, according to the single-crystal X-ray data, the complex has a different (2:1) composition featuring the sandwich structure. The complexes of ferrocene with copper and silver macrocycles have a columnar structure (X-ray data).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
481718
Iron(0) pentacarbonyl, >99.99% trace metals basis
C5FeO5
195731
Iron(0) pentacarbonyl
C5FeO5