EMAIL THIS PAGE TO A FRIEND
Expert review of molecular diagnostics
Response to a third-party evaluation of Clinitest® hCG sensitivity to various isoforms of human chorionic gonadotropin.
PMID 21405962
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11945 Benedict’s reagent, for the qualitative determination of sugars