EMAIL THIS PAGE TO A FRIEND
Parasites & vectors
Oral dosing with papaya latex is an effective anthelmintic treatment for sheep infected with Haemonchus contortus.
PMID 21406090
Abstract
The cysteine proteinases in papaya latex have been shown to have potent anthelmintic properties in monogastric hosts such as rodents, pigs and humans, but this has not been demonstrated in ruminants. In two experiments, sheep were infected concurrently with 5,000 infective larvae of Haemonchus contortus and 10,000 infective larvae of Trichostrongylus colubriformis and were then treated with the supernatant from a suspension of papaya latex from day 28 to day 32 post-infection. Faecal egg counts were monitored from a week before treatment until the end of the experiment and worm burdens were assessed on day 35 post-infection. We found that the soluble fraction of papaya latex had a potent in vivo effect on the abomasal nematode H. contortus, but not on the small intestinal nematode T. colubriformis. This effect was dose-dependent and at tolerated levels of gavage with papaya latex (117 μmol of active papaya latex supernatant for 4 days), the H. contortus worm burdens were reduced by 98%. Repeated treatment, daily for 4 days, was more effective than a single dose, but efficacy was not enhanced by concurrent treatment with the antacid cimetidine. Our results provide support for the idea that cysteine proteinases derived from papaya latex may be developed into novel anthelmintics for the treatment of lumenal stages of gastro-intestinal nematode infections in sheep, particularly those parasitizing the abomasum.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
757349
Fluoroethylene carbonate, 99%
C3H3FO3