EMAIL THIS PAGE TO A FRIEND
Journal of cardiovascular medicine (Hagerstown, Md.)
Black aorta in a patient with alkaptonuria (ochronosis).
PMID 21415764
Abstract
A rare cause of valvular heart disease is the deposition of foreign material in the valvular tissues, including material accumulating as a result of inborn errors of metabolism of the essential amino acids. Alkaptonuria can result in accumulation of homogentisic acid. We report the case of a patient with alkaptonuria undergoing surgery for aortic valve replacement.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0751
Homogentisic acid, crystalline
C8H8O4