EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Improved synthesis of 15-deoxyspergualin analogs using the Ugi multi-component reaction.
PMID 21420295
Abstract
Spergualin is a natural product that exhibits immunosuppressive, anti-tumor and anti-bacterial activities. Its derivatives, such as 15-deoxyspergualin (15-DSG), have been clinically approved for acute allograft rejection. However, the reported syntheses are cumbersome (>10 steps) and they suffer from low overall yields (∼0.3% to 18%). Moreover, spergualin and its derivatives are chemically unstable and rapidly hydrolyzed in aqueous buffer. Here, we have re-explored these issues and report a modified synthetic route with significantly improved overall yield (∼31% to 47%). The key transformation is a microwave-accelerated Ugi multi-component reaction that is used to generate the peptoid core in a single step. Using the products of this route, we found that modifications of the hemiaminal significantly increased chemical stability. Thus, we anticipate that this synthetic route will improve access to biologically active 15-DSG derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5822 Spergualin trihydrochloride, >95% (HPLC and TLC), from Bacillus laterosporus
C17H40Cl3N7O4