EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design and SAR of macrocyclic Hsp90 inhibitors with increased metabolic stability and potent cell-proliferation activity.
PMID 21420297
Abstract
A novel series of macrocyclic ortho-aminobenzamide Hsp90 inhibitors is reported. A basic nitrogen within the tether linking the aniline nitrogen atom to a tetrahydroindolone moiety allowed access to compounds with good physical properties. Important structure-activity relationship information was obtained from this series which led to the discovery of a soluble and stable compound which is potent in an Hsp90 binding and cell-proliferation assay.