EMAIL THIS PAGE TO A FRIEND
Dental materials journal
Study on ceramic coating on the enamel surface using a carbon dioxide laser.
PMID 21422667
Abstract
The aims of this study were to evaluate a new restorative method using a carbon dioxide laser (CO(2)-laser) and to evaluate the acid resistance of teeth. Experimental calcium phosphate glass (CPG) powder and two low melting point ceramics (Finesse and zirconium silicate) were fused to enamel surfaces using a CO(2)-laser at an irradiation intensity of 1.0 watt for 30 seconds with a beam size of 0.49 mm at the focal point. The treated teeth were observed with a scanning electron microscope, and the acid resistance of the treated enamel surfaces was evaluated. The CPG fused successfully to the enamel surface, and the treated enamel surface showed high acid resistance compared with the low melting point ceramics and the non-irradiated surfaces. This system may lead to the development of new restorative methods that do not require the use of bonding agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
383287
Zirconium(IV) silicate, −325 mesh
O4SiZr