EMAIL THIS PAGE TO A FRIEND
Urologiia (Moscow, Russia : 1999)
[Characteristics of microbiological diagnosis of chronic bacterial prostatitis].
PMID 21427995
Abstract
We made a microbiological study of the urine and prostatic secretion (Meares-Stamey test) of 35 patients with chronic bacterial prostatitis (CBP) aged 35-75 years. We conducted a bacterial assay, determined species, sensitivity to synthetic antibacterial fluoroquinolone drugs. A total of 163 bacterial strains were isolated, of them 13 were staphylococcal. Gram-negative bacteria were identified in the test biomaterial only in 16.6% cases. The analysis of staphylococcal sensitivity showed the highest antistaphylococcal effectiveness of levofloxacin and lomefloxacin. Our study confirms an essential role of staphylococcal bacteria in CBP etiology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2906
Lomefloxacin hydrochloride
C17H19F2N3O3 · HCl