EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Synthesis of N-(aminobutyl)-N-(ethylisoluminol) functionalized gold nanomaterials for chemiluminescent bio-probe.
PMID 21431126
Abstract
A novel strategy for the synthesis of CF-AuNMs by reducing HAuCl(4) with ABEI in aqueous solution at room temperature is reported. No additional stabilizing and reducing reagents are needed, and various morphologies of CF-AuNMs can be obtained. It could be used as bio-probe for bioassays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0156
N-(4-Aminobutyl)-N-ethylisoluminol, ≥90%
C14H20N4O2