EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Addition of boranes to N-aryl-salicylaldimines: intramolecular hydrogenation of imines.
PMID 21431160
Abstract
Addition of boranes to N-aryl-salicylaldimines takes place initially at the reactive phenolic O-H bond to give an activated boron-containing imine and dihydrogen. In some cases a subsequent intramolecular hydrogenation step is observed and the C=N imine bond is reduced to the corresponding amine. Reactions with dimesitylborane in THF are unique in that the reduced amine product is the major product observed in solution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
84230
Salicylamide, puriss., ≥99.0% (T)
C7H7NO2
860417
Salicylamide, 99%
C7H7NO2