EMAIL THIS PAGE TO A FRIEND
Investigative Ophthalmology & Visual Science
Adult donor rod photoreceptors integrate into the mature mouse retina.
PMID 21436277
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB5585
Anti-Recoverin Antibody