EMAIL THIS PAGE TO A FRIEND
Stomatologiia
[The use of iodobromine water on patients with periodontitis at a health resort].
PMID 2143850
Abstract
The technique of iodobromine water irrigations of the oral cavity in periodontitis has been developed to be used at health resorts of Anapa. A total of 4650 patients aged 25 to 52 with periodontitis of varying severity were treated; mineral water irrigations were preceded by removal of dental deposit. The treatment efficacy was assessed by functional methods: electrothermometry and rheography.
Related Materials