EMAIL THIS PAGE TO A FRIEND
Nanoscale
Green fluorescent organic nanoparticles by self-assembly induced enhanced emission of a naphthalene diimide bolaamphiphile.
PMID 21445422
Abstract
Naphthalene diimide (NDI) bolaamphiphilic molecules (1) self-assemble in water to form organic nanoparticles, which exhibit self-assembly induced preassociated excimer formation and hence an enhanced green fluorescence.