EMAIL THIS PAGE TO A FRIEND
Cancer research
Combined treatment of pancreatic cancer with mithramycin a and tolfenamic Acid promotes sp1 degradation and synergistic antitumor activity--letter.
PMID 21447740
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34073
Tolfenamic acid, VETRANAL®, analytical standard
C14H12ClNO2