EMAIL THIS PAGE TO A FRIEND
Journal of magnetic resonance (San Diego, Calif. : 1997)
DEER in biological multispin-systems: a case study on the fatty acid binding to human serum albumin.
PMID 21450500
Abstract
In this study, self-assembled systems of human serum albumin (HSA) and spin-labeled fatty acids are characterized by double electron-electron resonance (DEER). HSA, being the most important transport protein of the human blood, is capable to host up to seven paramagnetic fatty acid derivatives. DEER measurements of these self-assembled multispin clusters are strongly affected by correlations of more than two spins, the evaluation of the latter constituting the central topic of this paper. While the DEER modulation depth can be used to obtain qualitative information of the number of coupled spins, the quantitative analysis is hampered by the occurrence of cluster mixtures with different numbers of coupled spins and contributions from unbound spin-labeled material. Applying flip angle dependent DEER measurements, unwanted multispin correlations were found to lead not only to a broadening of the distance peaks but also to cause small distances to be overestimated and large distances to be suppressed. It is thus favorable to use spin-diluted systems with an average of two paramagnetic molecules per spin cluster when a quantitative analysis of the distance distribution is sought.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
253596
16-DOXYL-stearic acid, free radical
C22H42NO4