EMAIL THIS PAGE TO A FRIEND
Journal of gastrointestinal and liver diseases : JGLD
Fluindione: a francophone oral anticoagulant drug utilized in liver transplant recipients for Budd-Chiari syndrome.
PMID 21451810
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P26406
2-Phenyl-1,3-indandione, 97%
C15H10O2