EMAIL THIS PAGE TO A FRIEND
Magnetic resonance in chemistry : MRC
Complete NMR data of methoxylated cis- and trans-stilbenes as well as 1,2-diphenylethanes.
PMID 21452348
Abstract
Resveratrol is a polyphenol isolated from many natural sources including grapes, mulberries, eucalyptus, spruce, lilies, and peanuts. The hydroxyl groups in polyphenols can be substituted with various functional groups, allowing production of multiple derivatives. NMR spectroscopy is used to identify new derivatives. Since the complete NMR data of the known derivatives can be useful for identification of the newly isolated derivatives, here, we report the synthesis of 14 methoxylated stilbenes and four 1,2-diphenylethanes and their NMR data.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B33706
Bibenzyl, ReagentPlus®, 99%
C14H14