EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Determination of phytic acid and inositolphosphates in barley.
PMID 21455911
Abstract
Phytic acid (PA) and lower inositolphosphates (InsP(n) ) is the main storage form of phosphorus in grains or seeds. The content of PA and InsP(n) in different varieties of barley was analyzed by capillary isotachophoresis and online-coupled capillary isotachophoresis with CZE. The electrolytes (in demineralized water) for the isotachophoretic analysis consisted of 10 mM HCl, 14 mM glycylglycine, and 0.1% 2-hydroxyethylcellulose (leading) and 10 mM citric acid (terminating). The optimized electrolytes for the online coupling isotachophoresis with zone electrophoresis analysis were mixtures of 5 mM HCl, 7 mM glycylglycine, and 0.1% 2-hydroxyethylcellulose (leading), 20 mM citric acid, 10 mM glycylglycine, and 0.1% 2-hydroxyethylcellulose (background) and 10 mM citric acid (terminating). PA and all studied InsP(n) were separated within 25 min and detected by a conductivity detector. Simple sample preparation (acidic extraction), sufficient sensitivity, speed of analysis, and low running cost are important attributes of the electrophoretic methods. The method was used for the determination of PA and InsP(n) in barley varieties within an ongoing research project.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1127
Gly-Gly hydrochloride
C4H8N2O3 · HCl