EMAIL THIS PAGE TO A FRIEND
The Journal of eukaryotic microbiology
Balamuthia mandrillaris: staining properties of cysts and trophozoites and the effect of 2,6-dichlorobenzonitrile and calcofluor white on encystment.
PMID 21462548
Abstract
Here, we determined the staining properties of Balamuthia mandrillaris cysts, and assessed the effect of 2, 6-dichlorobenzonitrile (DCB), a cellulose synthesis inhibitor, and calcofluor white, a brightening agent, on its encystment. Periodic acid-Schiff reagent stained the inner wall intensely and middle and outer walls weakly suggesting that the cyst wall of B. mandrillaris may contain glycans. Furthermore, cysts, but not trophozoites, fluoresced when stained with calcofluor white. Calcofluor white and DCB, a cellulose synthesis inhibitor, inhibited B. mandrillaris encystment. This is the first report suggesting possible glycan biosynthesis in B. mandrillaris encystment, and this pathwaymay provide a potentially useful drug target and help improve treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D57558
2,6-Dichlorobenzonitrile, 97%
C7H3Cl2N
45431
Dichlobenil, PESTANAL®, analytical standard
C7H3Cl2N