EMAIL THIS PAGE TO A FRIEND
Organic letters
Diasteroselective preparation of cyclopropanols using methylene bis(iodozinc).
PMID 21462964
Abstract
A diastereoselective synthesis of trans-2-substituted cyclopropanols is outlined. Bimetallic CH(2)(ZnI)(2) was found to react with α-chloroaldehydes to give cyclopropanols in yields of 64-89% and dr's ≥ 10:1. The high trans-selectivity resulted from equilibration of the cyclopropoxide intermediates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
158429
Diiodomethane, ReagentPlus®, 99%, contains copper as stabilizer
CH2I2