EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and biological evaluation of benzyl styrylsulfonyl derivatives as potent anticancer mitotic inhibitors.
PMID 21463944
Abstract
We herein report the synthesis, biological activity and structure activity relationship of derivatives of benzylstyrylsulfone, benzylstyrylsulfine and benzylsulfonyl-N-phenylacetamide. A lead compound 7 represents a new class of mitotic inhibitors that demonstrates potent anti-proliferative activity and selectively induces cancer cell apoptosis while sparing non-transformed lung fibroblast.
Related Materials