EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Bioluminescence imaging evaluation of the inhibitory effect of lidamycin on lung metastasis of human fibrosarcoma in athymic mice].
PMID 21465808
Abstract
This study is to investigate the inhibitory effect of lidamycin (LDM) and its combination with methotrexate (MTX) on lung metastasis of fibrosarcoma by bioluminescence imaging in athymic mice. A stable luciferase transfected HT-1080 cell line was constructed and the capability to establish experimental lung metastasis in athymic mice was confirmed. The optical imaging system was applied to evaluate the formation of lung metastasis in vivo. In addition, metastatic nodules were counted for the evaluation of inhibition rates. As shown, the fluorescent intensity of luciferase-transfected HT-1080 cells was colinear with the cell population and the minimal detected cell population was 100 cells/well. Optical imaging showed that the fluorescent intensity of treated group was apparently lower than that of the control. The inhibition rates of lung metastasis by LDM alone at 0.025 mg x kg(-1) and 0.05 mg x kg(-1) were 53.9% and 75.9%, respectively, while that of MTX alone at 0.5 mg x kg(-1) was 70.2%. The combination of LDM at 0.025 mg x kg(-1) and MTX at 0.5 mg x kg(-1) showed an inhibition rate of 88.7%. The coefficient of drug interaction (CDI) was 0.82. The results herein demonstrated that LDM alone had strong anti-metastasis effect on human fibrosarcoma HT-1080 and the inhibition efficacy is strengthened when combined with MTX.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
HT 1080