EMAIL THIS PAGE TO A FRIEND
Rhinology
Paradoxical increase in nasal airway resistance following a topical nasal decongestant spray.
PMID 21468388
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X6000
Xylometazoline hydrochloride
C16H24N2 · HCl