EMAIL THIS PAGE TO A FRIEND
Chimia
Syntheses and biological activities of 1,4-iminoalditol derivatives as alpha-L-fucosidase inhibitors.
PMID 21469443
Abstract
A review dealing with 1,4-iminoalditol (hydroxylated pyrrolidine) derivatives as inhibitors of alpha-L-fucosidases including the different synthetic approaches for their preparation as well as their inhibitory properties is presented.
Related Materials