EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Water-mediated conformations of the alanine dipeptide as revealed by distributed umbrella sampling simulations, quantum mechanics based calculations, and experimental data.
PMID 21469661
Abstract
An exhaustive umbrella sampling simulation of the alanine dipeptide in solution is reported. The presence of stable Y conformations in solution is assessed by both quantum calculations and experimental data from X-ray and NMR protein-solved structures available in the protein coil library. The agreement between experimental and simulation Ramachandran plots of the dipeptide in solution is excellent. A suitable explanation of the stabilization of the Y conformation mediated by water for the alanine dipeptide is discussed on the basis of Car-Parrinello MD calculations of the dipeptide in water.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0878
Ala-Gly
C5H10N2O3
A1878
Ala-Leu
C9H18N2O3
A2003
Ala-Lys hydrochloride
C9H19N3O3 · HCl
A3128
Ala-Phe
C12H16N2O3
A3253
Ala-Pro hydrate
C8H14N2O3 · xH2O
A4003
Ala-Tyr
C12H16N2O4