EMAIL THIS PAGE TO A FRIEND
Clinical nutrition (Edinburgh, Scotland)
Additional data from our study on fatty acids variations during lactation: correlations between n-3 and n-6 PUFAs in human colostrum, transitional, and mature milk.
PMID 21470725
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1797
all-cis-7,10,13,16,19-Docosapentaenoic acid, synthetic, ≥97%
C22H34O2