EMAIL THIS PAGE TO A FRIEND
Natural product reports
Muscarine, imidazole, oxazole, and thiazole alkaloids.
PMID 21472175
Abstract
A great number of structurally diverse natural products containing five-membered heterocyclic subunits, such as imidazole, oxazole, thiazole, and their saturated congeners, are abundant in nature. These naturally occurring metabolites often exhibit extensive and pharmacologically important biological activities. The latest progress in the isolation, biological activities, chemical synthetic studies, and biosynthetic pathways on these natural products is summarized in this review.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2399
Imidazole, ACS reagent, ≥99% (titration)
C3H4N2