EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Stereoselectivity of isolated dehydratase domains of the borrelidin polyketide synthase: implications for cis double bond formation.
PMID 21472957
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3061
Borrelidin, from Streptomyces parvulus, ≥98% (HPLC)
C28H43NO6