EMAIL THIS PAGE TO A FRIEND
Pest management science
Dynamic plant uptake model applied for drip irrigation of an insecticide to pepper fruit plants.
PMID 21472970
Abstract
Drip application of insecticides is an effective way to deliver the chemical to the plant that avoids off-site movement via spray drift and minimizes applicator exposure. The aim of this paper is to present a cascade model for the uptake of pesticide into plants following drip irrigation, its application for a soil-applied insecticide and a sensitivity analysis of the model parameters. The model predicted the measured increase and decline of residues following two soil applications of an insecticide to peppers, with an absolute error between model and measurement ranging from 0.002 to 0.034 mg kg fw(-1). Maximum measured concentrations in pepper fruit were approximately 0.22 mg kg fw(-1). Temperature was the most sensitive component for predicting the peak and final concentration in pepper fruit, through its influence on soil and plant degradation rates. Repeated simulations of pulse inputs with the cascade model adequately describe soil pesticide applications to an actual cropped system and reasonably mimic it. The model has the potential to be used for the optimization of practical features, such as application rates and waiting times between applications and before harvest, through the integrated accounting of soil, plant and environmental influences.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36159
Methomyl, PESTANAL®, analytical standard
C5H10N2O2S