EMAIL THIS PAGE TO A FRIEND
Leukemia research
Dequalinium induces human leukemia cell death by affecting the redox balance.
PMID 21477862
Abstract
Dequalinium, an amphiphilic quinolinium derivative, selectively accumulates in mitochondria and displays anticancer activity in cells from different malignancies. Previous studies indicate a differential DQA-induced cytotoxicity in NB4 and K562 human leukemia cells as a consequence of an early disturbance in mitochondrial function. Results in this paper show that DQA induces a concentration-dependent oxidative stress by decreasing GSH level and increasing ROS in a cell type specific way. Inhibitors of the JNK and p38 stress regulated kinases potentiate DQA-induced NB4 cell death suggesting a protective function for these enzymes. K562 cells with relatively high GSH levels remained resistant to DQA action.