EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
New solid phase synthesis of distamycin analogues.
PMID 21483272
Abstract
A novel and straightforward solid phase synthesis of distamycin analogues containing benzene units has been developed.