EMAIL THIS PAGE TO A FRIEND
Cardiovascular drugs and therapy
Istaroxime: is the remedy better than the disease?: Editorial to: "Chronic istaroxine improves cardiac function and heart rate variability in cardiomyopathic hamsters" by Pietro Lo Giudice et al.
PMID 21484265
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E5126
Etiocholan-3α-ol-17-one
C19H30O2