EMAIL THIS PAGE TO A FRIEND
ACS chemical biology
Carba-sugars activate the glmS-riboswitch of Staphylococcus aureus.
PMID 21486059
Abstract
The glmS-riboswitch is unique among riboswitch families as it represents a metabolite-dependent ribozyme that undergoes self-cleavage upon recognition of glucosamin-6-phosphate. The glmS-riboswitch is located in the 5'-untranslated region of bacterial genes involved in cell wall biosynthesis. Therefore, this riboswitch represents a promising target for developing new antibiotics. We describe the metabolite-dependent glmS-riboswitch of pathologically relevant and vancomycin-resistant Staphylococcus aureus and the discovery and synthesis of a carba-sugar with potency similar to that of the native metabolite glucosamine-6-phosphate in modulating riboswitch activity. This compound represents a valuable lead structure for the development of antibiotics with a novel mode of action.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5509
D-Glucosamine 6-phosphate, ≥98%
C6H14NO8P
G4878
D-Glucosamine 6-phosphate sodium salt, ≥98%
C6H14NO8P