EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Highly enhanced fluorescence of fluorophores inside a metallic nanocavity.
PMID 21487583
Abstract
Here we report the observation of significantly enhanced fluorescence from molecules placed inside a silver nanocavity, as compared to those without the silver coating.