EMAIL THIS PAGE TO A FRIEND
The protein journal
Mercuric ion stabilizes levansucrase secreted by Acetobacter nitrogenifigens strain RG1(T).
PMID 21487805
Abstract
The purification and characterization of an extracellular levansucrase enzyme produced by novel nitrogen-fixer Acetobacter nitrogenifigens strain RG1(T) is described. Culture conditions were optimized for maximum levansucrase production. Levansucrase purified to homogeneity by tenfold purification has a molecular weight of 65 kDa, contained four cysteine residues, polymerized raffinose and was stable for 21 days at pH 6.0 when stored at 4 °C or -20 °C but was vulnerable to DTT and β-mercaptoethanol. Interestingly, this enzyme showed enhanced hydrolytic and polymerization activity in the presence of mercuric ion which, to our knowledge, is the first report for any levansucrase enzyme characterized so far. Evidences obtained from Native PAGE, tryptophan fluorescence study and activity measurements at different temperatures and in the presence of thiol modifying agents, show that mercuric ion stabilizes the enzyme. Levan, synthesized by the enzyme, has a molecular weight of 7,080 kDa and was shown to be a homopolymer of fructose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L8647 Levan from Erwinia herbicola